Learn More
MilliporeSigma™ LiChroCART™ HPLC Cartridge, 250-4.6
Catalog No. p-7111342 Shop All MilliporeSigma Products
Change view
Click to view available options
Stationary Phase:
RP18
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Stationary Phase
M1514317138 RP18
M1514317091 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1514317138 Supplier MilliporeSigma™ Supplier No. 1514317138
Only null left
Add to Cart
Add to Cart
Available in a variety of stationary phases that function in normal and/or reverse phase. MilliporeSigma LiChroCART™ HPLC Cartridge, 250-4.6 comes with customized packing (please specify packing material) and is 250mmL x 4.6mmI.D.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 4.6 mm
Length (Metric) 250 mm
Mobile Phase Compatibility RP (reversed-phase)
Stationary Phase RP18
Packing Material Customized (specify packing material)
Column Type LiChroCART™ HPLC Cartridge