Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Benzylamine, For GC Derivatization, 99.0%, MilliporeSigma™ Supelco™

Catalog No. 111020458 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0458 100 mL
11-101-9323 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-0458 Supplier MilliporeSigma Supelco Supplier No. 13180100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 100-46-9
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.16
MDL Number MFCD00008106
InChI Key WGQKYBSKWIADBV-UHFFFAOYSA-N
Synonym alpha-Aminotoluene
IUPAC Name 1-phenylmethanamine
SMILES NCC1=CC=CC=C1

Specifications

Melting Point 10°C
Density 0.981 g/mL (at 25°C)
Boiling Point 184°C to 185°C
Refractive Index n20/D 1.543; n20/D 1.543
Linear Formula C6H5CH2NH2
Quantity 100 mL
Formula Weight 107.15
Percent Purity ≥99.0% (GC)
Grade For GC derivatization
Product Line LiChropur
RTECSNumber DP1488500

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.