Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LiChropur™ Ethylenediaminetetraacetic acid disodium salt dihydrate, 99.0-101.0% (KT), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
10 g
50 g
Specifications
Specifications
Melting Point | 248°C |
CAS | 6381-92-6 |
Quantity | 10 g |
Linear Formula | C10H14N2 Na2O8 · 2H2 O |
Solubility Information | Water: 0.1 M (at 20°C, clear, colorless) |
Formula Weight | 372.24 |
Percent Purity | 99.0 to 101.0% (KT) |
Grade | HPLC |
Product Line | LiChropur |
Safety and Handling
RTECSNumber | AH4410000 |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction