Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Phosphoric Acid, MilliporeSigma™ Supelco™

Catalog No. 111022597 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2597 100 mL
11-102-2598 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-2597 Supplier MilliporeSigma Supelco Supplier No. 49685100ML
Add to Cart
Add to Cart

For HPLC

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.99
MDL Number MFCD00011340
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym Orthophosphoric Acid
IUPAC Name phosphoric acid
SMILES OP(O)(O)=O

Specifications

Melting Point ∼40°C (literature)
Boiling Point 158°C (literature)
Density 1.685 g/mL at 25°C (literature)
Vapor Pressure 2.2 mmHg (20°C)
Quantity 100 mL
Linear Formula H3PO4
Formula Weight 98
Percent Purity 85%
Grade HPLC
Hazard Category H290 - H314
RTECSNumber TB6300000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.