Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Sodium Phosphate Monobasic, MilliporeSigma™ Supelco™

Catalog No. 111022631 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2631 250 g
11-102-2632 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-2631 Supplier MilliporeSigma Supelco Supplier No. 52074250G
Add to Cart
Add to Cart

For HPLC

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-80-7

Specifications

pH Range 4 to 4.5 (20°C, 10% in H2O)
Purity or Quality Grade 99% to 101% (T)
Quantity 250 g

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.