Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Licorice Extract
SCI_ED

Catalog No. S25740 Shop All Fisherbrand Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Powder

This product is for educational use - no C of A is available.

Chemical Identifiers

Synonym Glycyrrhiza Extract

Specifications

Color Brown
Packaging Powder
Linear Formula C42H62O16
Quantity 25 g
Chemical Name or Material Licorice Extract
ShelfLife 2 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.