Learn More
Lide Labs Hansel™ Stains
Catalog No. 23233800
Encompass
Change view
Click to view available options
Quantity:
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-233-800 125 mL
23-233-801 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-233-800 Supplier Lide Labs Supplier No. HANSEL40Z
Only null left
Add to Cart
Add to Cart
For the examination of the nasal secretions to differentiate an allergic condition from a bacterial infection
  • One-minute stain
  • For secretion cytology
  • Stain eosinophils red; neutrophils, epithelial (exfoliated cells) and bacteria blue; and mast cells or basophiles red and blue
  • Can be adapted for staining various secretions: urinary, conjunctival, salivary, gastrointestinal, nasal, sinus, aural and bronchial
  • Determine elevated levels of eosinophils in urine (eosinophiluria) which can aid in diagnosis of many associated clinical conditions
  • Cannot be used for blood staining
Specifications
Type Hansel Stain
For Use With (Application) Secretion cytology stains eosinophils red, neutrophils, epithelial (exfoliated cells) and bacteria blue, and mast cells or basophiles red and blue
Quantity 125 mL