Learn More
Invitrogen™ Lidocaine Monoclonal Antibody (LC10-136.4)

Description
MA1-10751 detects Lidocaine. MA1-10751 may be used in immunoassays to detect and quantitate lidocane. The MA1-10751 immunogen is Lidocaine, conjugated to KLH.
Specifications
Specifications
| Antigen | Lidocaine |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | LC10-136.4 |
| Concentration | 1.27 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene Alias | Anestacon; C14H22N2O; Lidoderm; Lignocaine; Xylocaine |
| Host Species | Mouse |
| Immunogen | Lidocaine, conjugated to KLH. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.