Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lidocaine Monoclonal Antibody (LC11-378.5), QED Bioscience™
SDP

Catalog No. 502277729
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7729 100 μg
50-227-7730 500 μg
50-227-7728 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-227-7729 Supplier Leinco Technologies Inc Supplier No. 16604100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lidocaine Monoclonal antibody specifically detects Lidocaine in Chemical samples. It is validated for ELISA

Specifications

Antigen Lidocaine
Applications ELISA
Classification Monoclonal
Clone LC11-378.5
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias Anestacon; C14H22N2O; Lidoderm; Lignocaine; Xylocaine
Host Species Mouse
Immunogen Lidocaine-KLH.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.