Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lidocaine, USP, 97.5-102.5%, Spectrum™ Chemical
SDP

Catalog No. 18603877 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

LI102, 137-58-6, C14H22N2O

Spectrum™ Chemical Lidocaine, USP , also known as xylocaine or lignocaine is used as a topical local anesthetic and medication to numb tissue in a specific area. It also has uses as a treatment for ventricular tachycardia and to perform nerve blocks. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 137-58-6
Molecular Formula C14H22N2O
Molecular Weight (g/mol) 234.34
InChI Key NNJVILVZKWQKPM-UHFFFAOYSA-N
IUPAC Name 2-(diethylamino)-N-(2,6-dimethylphenyl)acetamide
SMILES CCN(CC)CC(=O)NC1=C(C)C=CC=C1C

Specifications

CAS Max % 100%
Quantity 25 kg
Packaging Fiber Drum
Percent Purity 97.5 to 102.5%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.