Learn More
United Scientific Supplies Liebig Condensers, 24/40 Joint
Shop All United Scientific Supplies, Inc Products
Description
- Use as part of a distillation apparatus
- Condenser has a straight inner tube that the condensing vapor passes through
- Cooling water passes through a larger outer jacket
- Socket is 24/40 with a screw thread connector
Specifications
Specifications
| Standard Taper Joint No | 24/40 |
| Length (Metric) Jacket | 300 mm |
| Material | Borosilicate Glass |
| For Use With (Application) | Cooling and Condensing Hot Vapor |
| For Use With (Equipment) | Distillation Glassware |
| Type | Liebig Condenser |