Learn More
United Scientific Supplies Liebig Condensers, 24/40 Joint
SDPSCI_ED
Catalog No. S43791 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Length (Metric) Jacket:
200 mm
300 mm
400 mm
500 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Jacket
S43791 500 mm
S43788 200 mm
S43789 300 mm
S43790 400 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S43791 Supplier United Scientific Supplies Supplier No. CNL500
Add to Cart
Add to Cart
Used to Cool and Condense Hot Vapor
  • Use as part of a distillation apparatus
  • Condenser has a straight inner tube that the condensing vapor passes through
  • Cooling water passes through a larger outer jacket
  • Socket is 24/40 with a screw thread connector
Specifications
Standard Taper Joint No 24/40
Length (Metric) Jacket 500 mm
For Use With (Application) Cooling and Condensing Hot Vapor
For Use With (Equipment) Distillation Glassware
Material Borosilicate Glass
Type Liebig Condenser