Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIF Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176554
Description
LIF Polyclonal specifically detects LIF in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence, Block/Neutralize.Specifications
LIF | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI | |
Rabbit | |
25 kDa | |
0.1 mg | |
Cell Cycle and Replication, Stem Cell Lines | |
3976 | |
Human, Mouse, Rat | |
IgG |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Blocking Assay | |
1 mg/mL | |
Western Blot 1-4 ug/ml, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 20 ug/mL, Block/Neutralize | |
NP_002300 | |
LIF | |
Antibody was raised against a 16 amino acid synthetic peptide near the center of human LIF. The immunogen is located within amino acids 50 - 100 of LIF. Amino Acid Squence: LYYTAQGEPFPNNLDK | |
Affinity Purified | |
RUO | |
Primary | |
At least two isoforms of LIF are known to exist. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction