Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIF Antibody (MM0454-6H31), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211773H
Description
LIF Monoclonal specifically detects LIF in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
LIF | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
LIF | |
Human recombinant LIF | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0454-6H31 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI | |
Mouse | |
Protein G purified | |
Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Lines | |
3976 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction