Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LIF Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579601
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Intestine Tissue, Rat Brain Tissue, Rat Spleen Tissue, Rat Thymus Tissue, Mouse Brain Tissue, Mouse Kidney Tissue, Mouse Liver Tissue.
Leukemia inhibitory factor (LIF) is a 20 kDa protein that belongs to the IL-6 receptor family. It binds to a heterodimeric membrane receptor made up of a LIF-specific subunit, gp190 or LIFR, and the subunit gp130, which is shared with the other members of the IL-6 family. LIF expression has been observed in various tissues including thymus, lung, and neuronal tissue. LIF can be up-regulated by pro-inflammatory cytokines such as TNFα and IL-17, and elevated levels of LIF have been found in cases of rheumatoid arthritis, neural injury, systemic inflammation, and tuberculosis. LIF displays diverse biological effects, but is best known for its ability to inhibit the differentiation of embryonic stem cells in mice and contribute to stem cell self-renewal.Human and mouse LIF share 79% sequence homology and exhibit cross-species activity. However, LIF inhibition of stem cell differentiation appears to be mouse-specific.
Specifications
LIF | |
Polyclonal | |
Unconjugated | |
Lif | |
CDF; cholinergic differentiation factor; cholinergic neuronal differentiation factor; d factor; DIA; differentiation inhibitory activity; differentiation-inducing factor; differentiation-stimulating factor; Emfilermin; hepatocyte-stimulating factor III; HILDA; human interleukin in DA cells; leukemia inhibitory factor; leukemia inhibitory factor (cholinergic differentiation factor); LIF; Melanoma-derived LPL inhibitor; MLPLI; myeloid leukaemia inhibitory factor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16878, 60584 | |
-20°C | |
Lyophilized |
Western Blot, Immunocytochemistry | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
P09056, P17777 | |
Lif | |
A synthetic peptide corresponding to a sequence at the C-terminus of mouse LIF (175-194aa DKEAFQRKKLGCQLLGTYKQ). | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction