Learn More
Walter Products™ Life Size Skeleton Model with Muscles & Ligaments
SCI_ED
Catalog No. s81000
Click to view available options
Quantity:
1
This full-size skeleton is a great instructional aid for teaching anatomy. Carefully cast with natural color plastic and pelvis mounted on a metal stand with wheels.
  • Life-size human skeleton made of high quality medical PVC
  • Muscle origins (red) and insertions (blue) are accurately painted on the left side
  • Ligaments are represented on the right side
  • Skull has movable jaw, removable skull cap, suture lines and a full dentition
  • Both arms and legs are joined by connectors and can be moved to demonstrate natural movement around the joint, they can also be removed for further study
  • Comes pelvic mounted on a roller stand
Specifications
Product Type Model, Human Skeleton with Hand Painted Muscles and Ligaments
For Use With (Application) Education
Height (Metric) 170 cm
Includes Skeleton & roller stand & diagram
Material PVC
Quantity 1
Size Life-size
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.