Learn More
Invitrogen™ ReadyProbes™ Cell Viability Imaging Kit, Blue/Red
Description
Includes
3 x 2.5mL NucBlue Live, 3 x 2.5mL propidium iodide
ReadyProbes™ Cell Viability Imaging Kit, Red/Blue contains NucBlue™ Live reagent (Hoechst 33342) and propidium iodide. Just add 2 drops of each stable reagent (at room temperature) to 1mL of cell growth media, then determine viability by counting total vs dead cells. NucBlue™ Live reagent stains the nuclei of all the cells and can be detected using a standard DAPI filter. Propidium iodide stains only the nuclei of cells with compromised plasma membrane integrity and is detected with a standard TRITC/RFP (orange) filter set.
- NucBlue™ Live reagent: stains the nuclei of all cells; detected with a standard DAPI filter (excitation/emission maxima: 360/460nm)
- Propidium iodide: stains the nuclei of dead cells with compromised plasma membrane; detected with standard TRITC/RFP (orange) filter set (excitation/emission maxima: 535/617nm)
Suggestions for use
- NucBlue™ Live reagent and propidium iodide can be added directly to cells in full growth media or a compatible buffer solution.
- In most cases, 2 drops/mL and an incubation time of 5 to 30 minutes will give bright nuclear staining; however, optimization may be needed for some cell types, conditions, and applications. In such cases, simply add more or fewer drops until the optimal staining intensity is obtained.
Specifications
Specifications
| Cell Type | Mammalian Cells, Eukaryotic Cells |
| Color | Blue, Red |
| Content And Storage | Store at 2° to 8°C, or at room temperature. |
| Description | ReadyProbes Cell Viability Imaging Kit, Blue/Red |
| Detection Method | Fluorescence |
| For Use With (Application) | Viability Assay |
| For Use With (Equipment) | Fluorescence Microscope |
| Product Type | Cell Viability Imaging Kit |
| Dye Type | 3 x 2.5 mL NucBlue™ Live; 3 x 2.5 mL propidium iodide |
| Format | Dropper bottles |
| Show More |
Frequently Asked Questions (FAQs)
This is not recommended. The ReadyProbes reagents were developed for imaging applications whereas the Ready Flow reagents were optimized for flow cytometry.
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.