Learn More
LifeScan™ SureStep™ System
Click to view available options
:
Surestep Control Solutn 2vl/pk
Surestep Pro Controls 3/pk
Description
Includes
Kit Contents:
- SureStep™
- Meter
- 25 SureStep™
- Test Strips
- PENLET™
- II Automatic Blood Sampler
- 25 Lancets
- Logbook
- Two AAA Batteries
- Carrying Case
- Control Solution
- Owners Manual†
- Stores up to 150 test results
- Results in 15 seconds
- Glucose range: 0 to 500mg/dL; hematocrit range: Adult 25 to 60%
- Offers testing date/time stamp and QC event flag
- Extra-large display
- Ten-test memory
- Blue confirmation dot indicates adequate sample application
† Includes SureStep™ Meter, operating guide and quick reference manual
Specifications
Specifications
| Product Type | Test Strip |
| For Use With (Equipment) | Sure step system |
| Quantity | 3 |