Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIFR alpha Antibody (110), Alexa Fluor™ 405, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290580AF405
Description
LIFR alpha Monoclonal specifically detects LIFR alpha in Mouse samples. It is validated for ELISA.Specifications
LIFR alpha | |
Monoclonal | |
Alexa Fluor 405 | |
CD118, CD118 antigen, FLJ98106, FLJ99923, leukemia inhibitory factor receptor, leukemia inhibitory factor receptor alpha, LIF receptor, LIF-R, SJS2, STWS, SWS | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse LIFR alpha (NP_038612.1; Met 1-Ser 828). | |
0.1 mL | |
Immunology | |
3977 | |
Mouse | |
Purified |
ELISA | |
110 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human LIFR | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction