Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIFR alpha Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232070G
Description
LIFR alpha Polyclonal antibody specifically detects LIFR alpha in Human, Mouse samples. It is validated for Western BlotSpecifications
LIFR alpha | |
Polyclonal | |
Western Blot | |
CD118, CD118 antigen, FLJ98106, FLJ99923, leukemia inhibitory factor receptor, leukemia inhibitory factor receptor alpha, LIF receptor, LIF-R, SJS2, STWS, SWS | |
The immunogen this antibody was made to, maps to a region between residue 1047 to 1097 of human Leukemia Inhibitory Factor Receptor Alpha using the numbering given in entry NP_001121143.1 (GeneID 3977). | |
0.1 mL | |
Immunology | |
3977 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction