Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIFR alpha Antibody (MM0455-9B23), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211774IR
Description
LIFR alpha Monoclonal specifically detects LIFR alpha in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
LIFR alpha | |
Monoclonal | |
DyLight 755 | |
P42702 | |
LIFR | |
Human recombinant LIFR alpha (extracellular domain) | |
0.1 mL | |
Immunology | |
3977 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry | |
MM0455-9B23 | |
Western Blot, Flow Cytometry | |
CD118, CD118 antigen, FLJ98106, FLJ99923, leukemia inhibitory factor receptor, leukemia inhibitory factor receptor alpha, LIF receptor, LIF-R, SJS2, STWS, SWS | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction