Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIFR alpha Antibody (MM0455-9B23), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211774PEATT594
Description
LIFR alpha Monoclonal specifically detects LIFR alpha in Human samples. It is validated for Flow Cytometry.Specifications
LIFR alpha | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
LIFR | |
Human recombinant LIFR alpha (extracellular domain) | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Flow Cytometry | |
MM0455-9B23 | |
Flow Cytometry | |
CD118, CD118 antigen, FLJ98106, FLJ99923, leukemia inhibitory factor receptor, leukemia inhibitory factor receptor alpha, LIF receptor, LIF-R, SJS2, STWS, SWS | |
Mouse | |
Protein G purified | |
Immunology | |
3977.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction