Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIFR Monoclonal Antibody (5D3G12)
GREENER_CHOICE

Catalog No. FERMA548535
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548535 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548535 Supplier Invitrogen™ Supplier No. MA548535
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LIFR Monoclonal Antibody for Western Blot, Flow, ELISA

Signal-transducing molecule. May have a common pathway with IL6ST. The soluble form inhibits the biological activity of LIF by blocking its binding to receptors on target cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIFR
Applications ELISA, Flow Cytometry, Western Blot, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 5D3G12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Lifr
Gene Accession No. P42702
Gene Alias A230075M04Rik; AW061234; CD118; CD118 antigen; D-factor/LIF receptor; FLJ98106; FLJ99923; leukemia inhibitor factor receptor alpha-chain; leukemia inhibitory factor receptor; leukemia inhibitory factor receptor alpha; leukemia inhibitory factor receptor alpha chain; LIF; LIF R beta; LIF receptor; Lifr; LIF-R; SJS2; soluble differentiation-stimulating factor receptor; STWS; SWS
Gene Symbols Lifr
Host Species Mouse
Immunogen Purified recombinant fragment of human CD118 (AA: 45-188) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3977
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.