Learn More
Invitrogen™ LIFR Polyclonal Antibody

Description
LIF Receptor alpha Polyclonal Antibody detects endogenous levels of LIF Receptor alpha protein.
Specifications
Specifications
| Antigen | LIFR |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Lifr |
| Gene Accession No. | P42702 |
| Gene Alias | A230075M04Rik; AW061234; CD118; CD118 antigen; D-factor/LIF receptor; FLJ98106; FLJ99923; leukemia inhibitor factor receptor alpha-chain; leukemia inhibitory factor receptor; leukemia inhibitory factor receptor alpha; leukemia inhibitory factor receptor alpha chain; LIF; LIF R beta; LIF receptor; Lifr; LIF-R; SJS2; soluble differentiation-stimulating factor receptor; STWS; SWS |
| Gene Symbols | Lifr |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.