Learn More
Light and Moisture Meter
SCI_ED
Catalog No. S45149 Shop All Luster Leaf Products, Inc. Products
Change view
Click to view available options
For Use With (Application):
Measure soil moisture and light conditions
Catalog No. For Use With (Application)
S45149 Measure soil moisture and light conditions
1 options
This item is not returnable. View return policy
Catalog No. S45149 Supplier Luster Leaf Products, Inc. Supplier No. 1830
This item is not returnable. View return policy
Instantly measures soil moisture and light conditions — no batteries required.
  • Built-in light meter features a chromatic filter to determine that plants receive the correct quality and intensity of light
  • Packaging includes a guide to the light and water needs of hundreds of plants and vegetables
Specifications
Type Light and Moisture Meter
For Use With (Application) Measure soil moisture and light conditions