Learn More
BINDER™ Light Photometer
Shop All BINDER Inc. Products
Click to view available options
No. of Places:
25 measuring points
Description
- 25 measuring points on 3 measurement levels
- Spectral distribution of either 385 to 785 nm or 250 to 785 nm
- For illumination, irradiance, visible light, and UV-A (8012-9010 only)
Specifications
Specifications
Description | Light photometry - including certificate, illumination and irradiance for visible light, 25 measuring points on 3 measurement levels, as well as spectral distribution (385 - 785 nm) |
For Use With (Equipment) | BINDER Chambers KBF LQC 240, KBF LQC 720 |
Material | Steel |
No. of Places | 25 measuring points |
Type | Light Photometer |
For Use With (Application) | Illumination, Irradiance, Visible Light |
Includes | Certificate |