Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Light's Solution, Oxidation-Reduction Potential (ORP) Standard, Ricca Chemical
SDP

Catalog No. p-3251865
Click to view available options
Quantity:
500 mL
1 L

Specifications

Chemical Name or Material Light's Solution
Packaging Natural Poly Bottle
Name Note Oxidation-Reduction Potential (ORP) Standard
CAS Min % 82.0, 8.51, 3.97, 3.23
CAS Max % 86.0, 8.86, 4.13, 3.36
CAS 7664-93-9, 7783-83-7, 7783-85-9
Physical Form Liquid
Color Yellow
Melting Point Approximately 0°C
Quantity 500 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.