Learn More
Novus Biologicals™ Lightning-Link (R) Alkaline Phosphatase Animal Free Antibody Labeling Kit
Shop All R&D Systems Products

Specifications
Specifications
| Content And Storage | -20°C |
| Quantity | 1 mg |
| For Use With (Application) | ELISA |
| Product Type | Antibody Labeling Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.