Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lightning-Link (R) Alkaline Phosphatase Antibody Labeling Kit
SDP

Catalog No. 7020010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
30 μg
300 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-200-10 300 μg
70-200-05 100 μg
70-200-15 1 mg
70-200-30 30 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 70-200-10 Supplier Novus Biologicals™ Supplier No. 7020010
Add to Cart
Add to Cart

Applications: ELISA

Alkaline phosphatase (AP) is a hydrolyase enzyme that is frequently conjugated to antibodies for use in immunoassays. PNPP is a popular colorimetric substrate for AP; the product of the reaction, p-nitrophenol, can be read at 405nM. Fluorogenic substrates (e.g. 4-methylumbelliferyl phosphate; MUP) may also be employed.

Order Info

Quantity

Specifications

Quantity 300 μg
Conjugate Alkaline Phosphatase
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-AP Modifier reagent, 1 vial of LL-AP Quencher reagent
Product Type Alkaline Phosphatase Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.