Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lightning-Link (R) APC-Cy7 Antibody Labeling Kit
SDP

Catalog No. 7650010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 x 100 μg Reaction
3 x 0.01 mg Reaction
300 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-500-10 300 μg
76-500-05 1 x 100 μg Reaction
76-500-15 1 mg
76-500-30 3 x 0.01 mg Reaction
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 76-500-10 Supplier Novus Biologicals™ Supplier No. 7650010
Add to Cart
Add to Cart

Applications: Flow Cytometry

APC/Cy7 is a tandem conjugate. In the Lightning-Link APC/Cy7 the tandem is excited at 633nm, wavelength at which the APC has its maximum absorption. The excited APC functions then as an energy donor, where the energy is transferred via FRET (Fluorescence Resonance Energy Transfer) to the Cy7. The Cy7 emits then the energy received from the APC in the form of long wavelength light at 776nm.

Order Info

Quantity

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 300 μg
Conjugate APC/Cy7
Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher FD reagent
Product Type APC-Cy7 Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.