Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lightning-Link (R) Fluorescein Antibody Labeling Kit
SDP

Catalog No. 7070010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
30 μg
300 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-700-10 300 μg
70-700-30 30 μg
70-700-15 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 70-700-10 Supplier Novus Biologicals™ Supplier No. 7070010
Add to Cart
Add to Cart

Applications: Immunofluorescence

Fluorescein is a fluorophore widely used in biological analysis, with a pH dependent absorption and emission spectrum. In TBS LL-Fluorescein shows an absorption maximum at 494 nm and an emission maximum at 520 nm.

Order Info

Quantity

Specifications

Quantity 300 μg
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Conjugate FITC
Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-AP Modifier reagent, 1 vial of LL-AP Quencher reagent
Product Type Fluorescein Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.