Learn More
Description
Order Info
Quantity
Specifications
Specifications
| Quantity | 1 mg |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Conjugate | HRP |
| Kit Contents | Glass vial(s) of Lightning-Link™ mix (1, 3 or 5 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher reagent |
| Product Type | HRP Antibody Labeling Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
