Learn More
Description
The antibody is covalently bonded to the label in a directional and controlled process at near-neutral pH. In view of the large size of the PE-Cy5 label, the amount of antibody used in a labeling reaction with the tandem must always be less than the pack size of LL PE-Cy5, in order that the PE-Cy5 is in a slight molar excess. The best ratio for any new antibody reagent must be determined by experimentation but 50-60ug of IgG antibody for every 100ug of LL PE-Cy5 usually gives optimal results. 50ug of antibody corresponds to an Ab:PE-Cy5 molar ratio of 1:1. The amount of antibody used for labeling should be the same as the pack size of LL-PE-Cy5. For any new antibody, optimization of the ratio of antibody to dye is often worthwhile. If your antibody needs to be concentrated, purified, or cleaned up, consider purchasing an antibody clean up kit.
Order Info
Quantity
Specifications
Specifications
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Quantity | 600 μg |
| Conjugate | PE/Cy5 |
| Kit Contents | Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher FD reagent |
| Product Type | PE-Cy5 Antibody Labeling Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
