Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lightning-Link (R) PE-Texas Red Antibody Labeling Kit
SDP

Catalog No. 7670030 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
180 μg
30 μg
60 μg
600 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-700-30 30 μg
76-700-05 60 μg
76-700-10 180 μg
76-700-15 600 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 76-700-30 Supplier Novus Biologicals™ Supplier No. 7670030

Antibody labeling kit

Lightning-Link is the easiest and quickest method available for making antibody (protein) conjugates, requiring just 30 seconds hands-on time. The antibody is covalently bonded to the label in a directional and controlled process at near-neutral pH.
  • The Lightning-Link conjugation kit allows PE-Texas Red tandem conjugations to set up in seconds, simply by adding a solution of the antibody to be labeled to a proprietary lyophilised mixture containing PE-Texas Red.
  • The amount of antibody used for labeling should be the same as the pack size of LL-PE-Texas Red. For any new antibody, optimization of the ratio of antibody to dye is often worthwhile.
  • If your antibody needs to be concentrated, purified, or cleaned up, consider purchasing an antibody clean up kit.


PE/TEXAS RED is a tandem conjugate. The PE is excited at 488nm and functions as an energy donor for the TEXAS RED. Energy is transferred from the PE to the TEXAS RED via energy resonance transfer. The TEXAS RED emits the energy received from the PE in the form of long wavelength light at 615nm.

Order Info

Quantity

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 30 μg
Conjugate PE/Texas Red
Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher FD reagent
Product Type PE-Texas Red Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.