Learn More
Novus Biologicals™ Lightning-Link (R) Rapid Alexa Fluor 555 Antibody Labeling Kit
SDP
Catalog No. p-7146012
Click to view available options
Quantity:
1 mg
100 μg
30 μg
300 μg
Antibody labeling kit
Lightning-Link™ Rapid Antibody Labeling kits enable direct labeling of antibodies, proteins, peptides or other biomolecules and allow conjugations to be set up in seconds, and used within minutes (less than 20 minutes).
Kit Contents:
  • 1 vial of LL Rapid Modifier reagent
  • 1 vial of LL Rapid Quencher reagent
  • Lightning-Link™ Rapid mix
Specifications
Quantity 100 μg
Product Type Antibody Labeling Kit
Content And Storage -20°C
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.