Learn More
Novus Biologicals™ Lightning-Link (R) Rapid Alexa Fluor 647 Antibody Labeling Kit
SDP
Catalog No. 3360010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
30 μg
300 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-600-10 300 μg
33-600-05 100 μg
33-600-15 1 mg
33-600-30 30 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 33-600-10 Supplier Novus Biologicals™ Supplier No. 3360010
Add to Cart
Add to Cart
Antibody labeling kit
Lightning-Link™ Rapid Antibody Labeling kits enable direct labeling of antibodies, proteins, peptides or other biomolecules and allow conjugations to be set up in seconds, and used within minutes (less than 20 minutes).
Kit Contents:
  • Lightning-Link™ Rapid mix
  • 1 vial of LL Rapid Quencher reagent
  • 1 vial of LL Rapid Modifier reagent
Specifications
Quantity 300 μg
Product Type Antibody Labeling Kit
Content And Storage -20°C