Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lightning-Link (R) Rapid AMCA Antibody Labeling Kit
SDP

Catalog No. 3130015 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
30 μg
300 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-300-15 1 mg
31-300-30 30 μg
31-300-10 300 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-300-15 Supplier Novus Biologicals™ Supplier No. 3130015

Antibody labeling kit

Unlike standard AMCA labeling procedures, where a large molar excess of AMCA is employed, the Lightning-Link AMCA kit combines a low molar ratio of AMCA:antibody and high labeling efficiency to eliminate the need for purification steps. AMCA (aminomethylcoumarin acetate) is a blue fluorescent dye commonly used in fluorescence microscopy, arrays and in-situ hybridisation. It has an excitation wavelength of 345-350nm and an emission wavelength of 440-460nm.

Order Info

Quantity

Specifications

Quantity 1 mg
Conjugate AMCA
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Kit Contents Glass vial(s) of Lightning-Link Rapid mix (1 or 3 vials, depending on pack size), 1 vial of Rapid Modifier reagent, 1 vial of Rapid Quencher reagent
Product Type Rapid AMCA Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.