Learn More
Novus Biologicals™ Lightning-Link (R) Rapid Atto633 Antibody Labeling Kit
Shop All R&D Systems Products

Description
Order Info
Quantity
Specifications
Specifications
| Conjugate | Atto 633 |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Quantity | 30 μg |
| For Use With (Application) | Flow Cytometry |
| Product Type | Rapid Atto633 Antibody Labeling Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.