Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lightning-Link (R) Rapid DyLight 633 Antibody Labeling Kit
SDP

Catalog No. 3250030 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
30 μg
300 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-500-30 30 μg
32-500-10 300 μg
32-500-15 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 32-500-30 Supplier Novus Biologicals™ Supplier No. 3250030

Antibody labeling kit

DyLight 594 generates red fluorescence for a wide array of fluorescence labeling-based applications. It has a strong absorption at 593nm, high fluorescence at 618nm (extinction coefficient 8.0 x10 cm-1M-1) and high quantum yield.
  • Lightning-Link™ Rapid technology can be used to label small quantities of antibody with 100% recovery
  • Easy to use antibody labeling kit with quick labeling times
  • From 10μg up to 2mg
  • Kit with 30 seconds hands-on time
  • No separation steps and no losses
  • Scalable technology
Applications:
  • Western Blotting, ELISA, Immunohistochemistry, Immunofluorescence and FACS analysis

Order Info

Quantity

Specifications

Conjugate DyLight 633
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 30 μg
Product Type Rapid DyLight 633 Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.