Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lightning-Link (R) Rapid DyLight 680 Antibody Labeling Kit
SDP

Catalog No. 3270010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
30 μg
300 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-700-10 300 μg
32-700-30 30 μg
32-700-15 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 32-700-10 Supplier Novus Biologicals™ Supplier No. 3270010
Add to Cart
Add to Cart

Antibody labeling kit

DyLight 680 provides red fluorescence for a wide array of fluorescence labeling-based applications. It has a strong absorption at 682nm, high fluorescence at 715nm (extinction coefficient 1.4 x10&superscript_begin;5&superscript_end;cm&superscript_begin;-1&superscript_end;M&superscript_begin;-1&superscript_end;) and high quantum yield.

Order Info

Quantity

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 300 μg
Conjugate DyLight 680
Product Type Rapid DyLight 680 Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.