Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lightning-Link (R) Rapid FluoProbes647H Antibody Labeling Kit
SDP

Catalog No. 3620010 Shop All R&D Systems Products
Change view
Click to view available options
Sufficient For:
1 x 2 mg Reaction
3 x 20 μg Reactions
3 x 200 μg Reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
36-200-10 3 x 200 μg Reactions
36-200-30 3 x 20 μg Reactions
36-200-15 1 x 2 mg Reaction
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 36-200-10 Supplier Novus Biologicals™ Supplier No. 3620010
Add to Cart
Add to Cart

Antibody labeling kit

Order Info

Quantity

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 30 μg
Sufficient For 3 x 200 μg Reactions
Product Type Rapid FluoProbes647H Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.