Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lightning-Link (R) Rapid Texas Red Antibody Labeling Kit
SDP

Catalog No. 3150015 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
30 μg
300 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-500-15 1 mg
31-500-30 30 μg
31-500-10 300 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-500-15 Supplier Novus Biologicals™ Supplier No. 3150015

Antibody labeling kit

TEXAS RED is a red fluorescent dye, used in histology, in fluorescence microscopy applications and in immunohistochemistry. It has a strong absorption at 595nm, with a high fluorescence at 615nm (extinction coefficient 8.0 x10&superscript_begin;4&superscript_end;cm&superscript_begin;-1&superscript_end;M&superscript_begin;-1&superscript_end;) and high quantum yield.

Order Info

Quantity

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 1 mg
Conjugate Texas Red
Kit Contents Glass vial(s) of Lightning-Link Rapid mix (1 or 3 vials, depending on pack size), 1 vial of Rapid Modifier reagent, 1 vial of Rapid Quencher reagent
Product Type Rapid Texas Red Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.