Learn More
Novus Biologicals™ Lightning-Link (R) Rapid Type A Biotin Antibody Labeling Kit
Shop All R&D Systems Products

Description
- Labels small quantities of antibody
- Circumvents desalting techniques that interupt protein labeling
- Biotinylated
- Minimises assay background
- Scalable
- Bond between label, biomolecule is covalent, therefore conjugate is stable
Specifications
Specifications
| For Use With (Application) | Labeling and detection |
| Quantity | 100 μg |
| Product Type | Antibody detection kit |
| Kit Contents | Lightning-Link Rapid Mix 1, LL-Rapid Modifier Reagent 1, LL-Rapid Quencher Reagent |
| Sufficient For | One reaction |
| Content And Storage | Store at −20°C. Avoid freeze-thaw cycles. |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.