Learn More
Novus Biologicals™ Lightning-Link (R) Rapid Type A Biotin Antibody Labeling Kit
Shop All R&D Systems Products

Description
- No desalting or dialysis steps
- Lightning-link Rapid technology can be used to label small quantities of antibody with 100% recovery
- Type A biotin kit is intended for assays in which a streptavidin-labeled detection reagent will be used.
Specifications
Specifications
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Quantity | 300 μg |
| Product Type | Antibody Labeling Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.