Learn More
Novus Biologicals™ Lightning-Link (R) Rapid Type B Biotin Antibody Labeling Kit
SDP
Catalog No. 3710010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
30 μg
300 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-100-10 300 μg
37-100-15 1 mg
37-100-30 30 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
For Research Use Only
Catalog No. 37-100-10 Supplier Novus Biologicals™ Supplier No. 3710010
Add to Cart
Add to Cart
For Research Use Only
Label antibodies, proteins, peptides and other biomolecules with biotin, using simple procedure. Set up conjugations in seconds, and use them within minutes.
  • No desalting or dialysis steps
  • Lightning-link Rapid technology can be used to label small quantities of antibody with 100% recovery
  • Type A biotin kit is intended for assays in which a streptavidin-labeled detection reagent will be used.
Specifications
Quantity 300 μg
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Type Antibody Labeling Kit