Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ LightShift™ EMSA Optimization and Control Kit

Catalog No. pi20148x
Click to view available options
Quantity:
100 Reactions

Identify and characterize protein-DNA binding interactions by EMSA with this extraordinarily robust and sensitive system.

Thermo Scientific™ LightShift Chemiluminescent EMSA Kit is an extraordinarily robust and sensitive system for performing electrophoretic mobility shift assays (EMSA) to identify and characterize protein-DNA binding interactions.

The kit includes reagents for setting up and customizing protein:DNA binding reactions, a control set of DNA and protein extract to test the kit system, stabilized streptavidin-HRP conjugate to probe for the biotin-labeled DNA target, and an exceptionally sensitive chemiluminescent substrate module for detection.The principle for LightShift™ EMSA Detection is similar to a Western blot. Biotin end-labeled duplex DNA is incubated with a nuclear extract or purified factor and electrophoresed on a native gel. The DNA is then rapidly (30 minutes) transferred to a positive nylon membrane, UV-crosslinked, probed with streptavidin-HRP conjugate and incubated with the substrate. The protocol from labeling to results can be accomplished in a single day. All you need to perform the assay are purified DNA target that has been end-labeled with biotin, the protein extract you wish to test, nylon membrane and basic electrophoresis equipment. DNA targets may be synthesized with 5' or 3' biotin labels or they may be labeled after synthesis using the Thermo Scientific Biotin 3' End DNA Labeling Kit (Part No. 89818). Nuclear, cytosolic or whole cell protein extracts may be obtained by a variety of methods, including the Thermo Scientific NE-PER Nuclear and Cytoplasmic Extraction Reagent Kit (Part No. 78833).

Highlights:

  • Includes EBNA control system to help new users develop a working assay and understand the methods used to confirm binding interaction specificity
  • Excellent for detecting low-abundance proteins in nuclear extracts
  • Sensitivity that surpasses radioactive and digoxigenin methods
  • Compatible with previously established binding conditions for popular DNA:protein interactions
TRUSTED_SUSTAINABILITY

Specifications

Assay EMSA Assay
Content And Storage Sufficient For: 100 binding reactions
• 10X Binding Buffer, 1 mL
• Biotin-EBNA Control DNA, 50 μL
• Unlabeled EBNA DNA, 50 μL
• EBNA Extract, 125 μL
• Poly (dI·dC), 125 μL
• 50% Glycerol, 500 μL
• 1% NP-40, 500 μL
• 1 M KCl, 1 mL
• 100mM MgCl2, 500 μL
• 200mM EDTA pH 8.0, 500 μL
• 5X Loading Buffer, 1 mL

Store at -20°C.
Format Kit
Includes 10X Binding Buffer (1 mL), Biotin-EBNA Control DNA (50 μL), Unlabeled EBNA DNA (50 μL), EBNA Extract (125 μL), Poly dI·dC (125 μL), 50% Glycerol (500 μL), 1% NP-40 (500 μL), 1M KCl (1 mL), 100mM MgCl(500 μL), 200mM EDTA pH 8.0 (500 μL), 5X Loading Buffer (1 mL)
For Use With (Equipment) myECL™ Imager, X-Ray Film
Product Line LightShift™
Product Type LightShift EMSA Optimization and Control Kit
Quantity 100 Reactions
Target Specificity Not Target-Specific
Technique Gel Shift
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.