Learn More
MSA™ Lightweight Aramid Fiber Welding Hood
Catalog No. 18139440 Shop All MSA Products
Change view
Click to view available options
Material:
Aramid Fiber
Catalog No. Material
18-139-440 Aramid Fiber
1 options
Catalog No. 18-139-440 Supplier MSA™ Supplier No. 486328
Add to Cart
Add to Cart
Requires
Fits over Welder's Adapter
Protects the welder's head, neck and shoulders from sparks
  • Rubber seal fits snugly over the Welder's Adapter
  • Helps protect cartridges when an air-purifying respirator is used in welding operations
Specifications
For Use With (Application) Welding
Material Aramid Fiber
Mounting Type Rubber seal fits snugly over the Welder's Adapter
Product Type Hood