Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LILRA1 Monoclonal Antibody (4A1E10A6)
GREENER_CHOICE

Catalog No. PIMA538557
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538557 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538557 Supplier Invitrogen™ Supplier No. MA538557
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody has been tested in indirect ELISA.

LILRA3 is a member of leukocyte immunoglobulin-like receptor gene family. Leukocyte Ig-like receptors (LIRs) are a family of inhibitory and stimulatory immunoreceptors expressed predominantly on monocytes and B cells and at lower levels on dendritic cells and natural killer (NK) cells. LILRA3 lacks a transmembrane region and may act as a soluble receptor for class I MHC antigens. It is expressed in B-cells, and at lower levels in natural killer (NK) cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LILRA1
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 4A1E10A6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene LILRA1
Gene Accession No. O75019
Gene Alias CD85 antigen-like family member I; CD85I; leucocyte Ig-like receptor A1; leukocyte immunoglobulin like receptor A1; leukocyte immunoglobulin-like receptor 6; Leukocyte immunoglobulin-like receptor subfamily A member 1; leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 1; LILRA1; LIR6; LIR-6
Gene Symbols LILRA1
Host Species Mouse
Immunogen Purified recombinant fragment of human LILRA1 (aa: extra 322-461) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11024
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.