Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LILRA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523966
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523966 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523966 Supplier Invitrogen™ Supplier No. PA523966
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LILRA2 Polyclonal Antibody for Western Blot, Flow

Leukocyte Ig-like receptors (LIRs) are a family of immunoreceptors expressed predominantly on monocytes and B cells and at lower levels on dendritic cells and natural killer (NK) cells. All LIRs in subfamily B have an inhibitory function (LILRB1; MIM 604811). LIRs in subfamily A, with short cytoplasmic domains lacking an immunoreceptor tyrosine-based inhibitory motif (ITIM) and with trans membrane regions containing a charged arginine residue, may initiate stimulatory cascades. One member of subfamily A (LILRA3; MIM 604818) lacks a transmembrane region and is presumed to be a soluble receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LILRA2
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene LILRA2
Gene Accession No. Q8N149
Gene Alias CD85 antigen-like family member H; CD85h; ILT1; ILT-1; Immunoglobulin-like transcript 1; leucocyte Ig-like receptor A2; leukocyte immunoglobulin like receptor A2; leukocyte immunoglobulin-like receptor 7; Leukocyte immunoglobulin-like receptor subfamily A member 2; leukocyte immunoglobulin-like receptor subfamily A member 2 soluble; leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 2; LILRA2; LIR7; LIR-7
Gene Symbols LILRA2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 55-81 amino acids from the central region of human LILRA2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11027
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.