Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LILRA3 Monoclonal Antibody (06)
GREENER_CHOICE

Catalog No. PIMA530645
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530645 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530645 Supplier Invitrogen™ Supplier No. MA530645
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human ILT6/LILRA3.

Leukocyte Ig-like receptors (LIRs) are a family of immunoreceptors expressed predominantly on monocytes and B cells and at lower levels on dendritic cells and natural killer (NK) cells. All LIRs in subfamily B have an inhibitory function (see, e.g., LILRB1, MIM 604811). LIRs in subfamily A, with short cytoplasmic domains lacking an immunoreceptor tyrosine-based inhibitory motif (ITIM) and with transmembrane regions containing a charged arginine residue, may initiate stimulatory cascades (see LILRA1, MIM 604810). One member of subfamily A (LILRA3) lacks a transmembrane region and is presumed to be a soluble receptor (Borges et al., 1997 [PubMed 9548455]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen LILRA3
Applications ELISA
Classification Monoclonal
Clone 06
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene LILRA3
Gene Accession No. Q8N6C8
Gene Alias CD85 antigen-like family member E; CD85e; HM31; HM43; ILT6; ILT-6; Immunoglobulin-like transcript 6; leucocyte Ig-like receptor A3; leukocyte immunoglobulin like receptor A3; leukocyte immunoglobulin-like receptor 4; leukocyte immunoglobulin-like receptor A3; leukocyte immunoglobulin-like receptor subfamily A member 3; leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member 3; LILRA3; LIR4; LIR-4; Monocyte inhibitory receptor HM43/HM31
Gene Symbols LILRA3
Host Species Mouse
Immunogen Recombinant Human ILT6/LILRA3 Protein (Met1-Glu439).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11026
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.